Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3641599 | Chr.18:3504024 - 3507639 on Build GRCh38 | Loss | DLGAP1 |
nsv1059696 | Chr.18:3494893 - 3572306 on Build GRCh38 | Gain | DLGAP1 |
dgv52n6 | Chr.18:3502646 - 3508570 on Build GRCh38 | Deletion | DLGAP1 |
dgv577e199 | Chr.18:3504050 - 3507531 on Build GRCh38 | Deletion | DLGAP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |