Assay Details
Target Gene Details
Entrez Gene ID: | 4137 |
Gene Name: | microtubule associated protein tau |
Gene Aliases: |
DDPAC, FTDP-17, MAPTL, MSTD, MTBT1, MTBT2, PPND, PPP1R103, TAU |
Location: |
Chr.17:45894382-46028334 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss | MAPT MGC57346-CRHR1 LOC644172 LRRC37A4P LRRC37A2 MAPT-IT1 STH LRRC37A ARL17B CRHR1 NSF NSFP1 ARL17A LOC107987254 LOC105376839 KANSL1-AS1 MGC57346 SPPL2C CRHR1-IT1 LOC107987253 MAPT-AS1 KANSL1 |
dgv3178n100 | Chr.17:45877004 - 46034461 on Build GRCh38 | Gain | MAPT MAPT-IT1 STH MAPT-AS1 KANSL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |