Assay Details
Target Gene Details
Entrez Gene ID: | 79922 |
Gene Name: | mitochondrial rRNA methyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.17:36601581-36634698 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRM1 | XM_011525275.2 | XP_011523577.1 | ||
XM_011525276.2 | XP_011523578.1 | |||
XM_017025112.1 | XP_016880601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833429 | Chr.17:36605619 - 36790120 on Build GRCh38 | Gain | MRM1 |
dgv3156n100 | Chr.17:36589879 - 36646211 on Build GRCh38 | Gain | MRM1 DHRS11 |
esv3640474 | Chr.17:36564700 - 36908342 on Build GRCh38 | Gain | LOC102723471 MRM1 DHRS11 GGNBP2 |
esv2758686 | Chr.17:36588163 - 36790120 on Build GRCh38 | Gain+Loss | MRM1 DHRS11 GGNBP2 |
esv3640475 | Chr.17:36602406 - 36633068 on Build GRCh38 | Gain | MRM1 |
nsv469760 | Chr.17:36519312 - 36683773 on Build GRCh38 | Loss | PIGW MYO19 MRM1 DHRS11 GGNBP2 |
nsv457736 | Chr.17:36609563 - 36634759 on Build GRCh38 | Gain | MRM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |