Assay Details
Target Gene Details
Entrez Gene ID: | 64788 |
Gene Name: | lipase maturation factor 1 |
Gene Aliases: |
C16orf26, HMFN1876, JFP11, TMEM112, TMEM112A |
Location: |
Chr.16:853632-981615 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMF1 | NM_022773.2 | NP_073610.2 | ||
NR_033645.1 | ||||
NR_033646.1 | ||||
XM_006720928.2 | XP_006720991.1 | |||
XM_006720929.3 | XP_006720992.1 | |||
XM_011522614.2 | XP_011520916.1 | |||
XM_011522615.1 | XP_011520917.1 | |||
XM_011522617.2 | XP_011520919.1 | |||
XM_011522618.2 | XP_011520920.1 | |||
XM_017023575.1 | XP_016879064.1 | |||
XM_017023576.1 | XP_016879065.1 | |||
XM_017023577.1 | XP_016879066.1 | |||
XM_017023578.1 | XP_016879067.1 | |||
XM_017023579.1 | XP_016879068.1 | |||
XM_017023580.1 | XP_016879069.1 | |||
AB075873.1 | BAD38655.1 | |||
AK022743.1 | BAB14218.1 | |||
AK025955.1 | BAB15295.1 | |||
AK093050.1 | ||||
AK125201.1 | ||||
AK225065.1 | ||||
AK225406.1 | ||||
BC010738.1 | AAH10738.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571046 | Chr.16:828324 - 1202441 on Build GRCh38 | Gain | SOX8 SSTR5-AS1 LMF1 CACNA1H SSTR5 LOC107984876 C1QTNF8 LMF1-AS1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | AMDHD2 RHBDF1 HBA2 PGP RAB26 MSLN RPS2 ARHGDIG CCDC78 FAM173A LOC100287175 HBZ CCDC154 SYNGR3 TPSG1 MAPK8IP3 CASKIN1 PDIA2 PRR35 HAGHL PRSS27 MRPL28 DNASE1L2 TELO2 CCNF PRSS29P LOC107984876 MSRB1 TPSD1 TPSB2 LINC00235 LOC105371047 SNORA10 CEMP1 TRAF7 LOC105371046 BAIAP3 LOC106660606 WDR24 HBQ1 GNG13 CLCN7 LMF1-AS1 DECR2 NPRL3 TMEM8A RPL23AP5 MEIOB CACNA1H SPSB3 TSR3 SSTR5 UNKL HBA1 WASIR2 TPSAB1 MIR6511B1 METRN PTX4 STUB1 MLST8 NHLRC4 NME3 NPW MIR5587 MIR940 MIR3177 MIR3677 FBXL16 NTHL1 HN1L MRPS34 NARFL POLR3K AXIN1 NOXO1 EME2 LOC105371038 RPUSD1 IFT140 PIGQ PDPK1 ABCA3 NTN3 PKD1 SNHG9 ATP6V0C SNORA78 MIR3176 SNORD60 E4F1 RAB40C KCTD5 MPG SSTR5-AS1 LOC101929280 WDR90 SLC9A3R2 TBC1D24 LOC107987233 LOC100134368 HAGH SOX8 HBM FAM234A LOC105371184 RAB11FIP3 BRICD5 GFER MIR6767 SNRNP25 FLJ42627 MIR6859-4 LUC7L LINC00254 SNHG19 NDUFB10 RNPS1 IGFALS MIR3180-5 SNORA64 ERVK13-1 ECI1 LOC105371049 GNPTG MIR3178 ZNF598 RHBDL1 WFIKKN1 FAHD1 LOC652276 UBE2I MIR1225 PRR25 CHTF18 CRAMP1 RNF151 RHOT2 MIR6768 C16orf59 JMJD8 ABCA17P TBL3 LOC729652 METTL26 TMEM204 CAPN15 MIR662 C1QTNF8 NME4 MIR4717 MCRIP2 LMF1 TSC2 C16orf91 LOC105371045 MIR4516 DDX11L10 HS3ST6 RPL3L RGS11 NUBP2 |
esv3637586 | Chr.16:798995 - 892120 on Build GRCh38 | Loss | LMF1 PRR25 GNG13 |
esv3637595 | Chr.16:859147 - 890552 on Build GRCh38 | Loss | LMF1 |
nsv1046349 | Chr.16:819378 - 942590 on Build GRCh38 | Loss | LMF1 LOC107984876 LMF1-AS1 |
nsv952898 | Chr.16:495001 - 1132900 on Build GRCh38 | Deletion | LOC105371038 RPUSD1 PIGQ WDR24 MSLN CCDC78 FAM173A GNG13 LMF1-AS1 MIR3176 LOC100287175 RHBDL1 WFIKKN1 RAB40C SSTR5-AS1 LOC101929280 PRR25 WDR90 SSTR5 CHTF18 PRR35 RHOT2 LOC107987233 JMJD8 HAGHL METRN SOX8 METTL26 STUB1 NHLRC4 CAPN15 LOC105371184 MIR5587 RAB11FIP3 LOC107984876 MIR662 C1QTNF8 FBXL16 MCRIP2 NARFL LMF1 LINC00235 |
esv2661544 | Chr.16:874758 - 881265 on Build GRCh38 | Deletion | LMF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |