Assay Details
Target Gene Details
Entrez Gene ID: | 23269 |
Gene Name: | MGA, MAX dimerization protein |
Gene Aliases: |
MAD5, MXD5 |
Location: |
Chr.15:41621139-41769943 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1049514 | Chr.15:41666284 - 41789012 on Build GRCh38 | Gain | MAPKBP1 MGA MIR626 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |