Assay Details
Target Gene Details
Entrez Gene ID: | 90527 |
Gene Name: | dual oxidase maturation factor 1 |
Gene Aliases: |
NIP, NUMBIP, mol |
Location: |
Chr.15:45117366-45129938 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv1002924 | Chr.15:45120086 - 45132601 on Build GRCh38 | Deletion |
![]() ![]() |
nsv569313 | Chr.15:45121343 - 45165260 on Build GRCh38 | Gain |
![]() ![]() |
esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)