Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOXL1 | NM_005576.3 | NP_005567.2 | ||
XM_011521555.1 | XP_011519857.1 | |||
AK303982.1 | ||||
AK314222.1 | ||||
BC015090.2 | AAH15090.1 | |||
BC068542.1 | AAH68542.1 | |||
L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion | LOXL1-AS1 TBC1D21 LOXL1 |
nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion | LOXL1-AS1 TBC1D21 LOXL1 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | PML GOLGA6A STOML1 LOXL1-AS1 LOC283731 LOC732265 ISLR2 LOXL1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | SCAMP2 BBS4 SEMA7A LOC732265 MIR4513 LOC101929333 GOLGA6A ISLR CYP1A1 CYP11A1 PPCDC LOXL1 LOC102723750 C15orf59-AS1 NPTN-IT1 REC114 CD276 STOML1 LOC283731 ARID3B ADPGK-AS1 LOC729739 GOLGA6C CCDC33 NPTN RPP25 CLK3 LMAN1L C15orf59 C15orf39 CSK MPI LOXL1-AS1 SCAMP5 ULK3 ISLR2 NEO1 ADPGK STRA6 EDC3 UBL7-AS1 TBC1D21 MIR6882 FAM219B HCN4 MIR6881 LOC105376731 UBL7 HIGD2B CPLX3 CYP1A2 PML COX5A LOC107984728 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |