Assay Details
Target Gene Details
Entrez Gene ID: | 8812 |
Gene Name: | cyclin K |
Gene Aliases: |
CPR4 |
Location: |
Chr.14:99481402-99511515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCNK | NM_001099402.1 | NP_001092872.1 | ||
XM_005268154.4 | XP_005268211.1 | |||
AK311075.1 | ||||
BC015935.1 | AAH15935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635481 | Chr.14:99277609 - 99600560 on Build GRCh38 | Gain | CCNK CCDC85C SETD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |