Assay Details
Target Gene Details
Entrez Gene ID: | 56339 |
Gene Name: | methyltransferase like 3 |
Gene Aliases: |
IME4, M6A, MT-A70, Spo8 |
Location: |
Chr.14:21498131-21511375 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
METTL3 | NM_019852.4 | NP_062826.2 | ||
XM_006720206.3 | XP_006720269.1 | |||
XM_011536968.1 | XP_011535270.1 | |||
AK000222.1 | ||||
AK300289.1 | ||||
AK304251.1 | ||||
AK304570.1 | ||||
BC001650.2 | AAH01650.1 | |||
BC003031.1 | AAH03031.1 | |||
BC052244.1 | AAH52244.1 | |||
DC295053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3633718 | Chr.14:21495207 - 21527525 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)