Assay Details
Target Gene Details
Entrez Gene ID: | 65125 |
Gene Name: | WNK lysine deficient protein kinase 1 |
Gene Aliases: |
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65 |
Location: |
Chr.12:752552-911452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain | LOC105369595 B4GALNT3 WNK1 LOC100049716 ERC1 RAD52 NINJ2 |
dgv2253n54 | Chr.12:741122 - 859323 on Build GRCh38 | Loss | WNK1 |
nsv556899 | Chr.12:741447 - 813242 on Build GRCh38 | Loss | WNK1 |
esv2760258 | Chr.12:759943 - 776028 on Build GRCh38 | Loss | WNK1 |
esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss | LOC105369595 B4GALNT3 LOC102723544 SLC6A13 KDM5A WNK1 CCDC77 LOC100049716 NINJ2 |
nsv556893 | Chr.12:628064 - 873155 on Build GRCh38 | Gain | WNK1 LOC100049716 NINJ2 |
nsv518946 | Chr.12:745165 - 852170 on Build GRCh38 | Loss | WNK1 |
dgv41n73 | Chr.12:757620 - 767334 on Build GRCh38 | Deletion | WNK1 |
dgv2256n54 | Chr.12:756804 - 769498 on Build GRCh38 | Loss | WNK1 |
nsv1159952 | Chr.12:764778 - 774012 on Build GRCh38 | Duplication | WNK1 |
dgv1336n100 | Chr.12:754667 - 767200 on Build GRCh38 | Loss | WNK1 |
dgv254e199 | Chr.12:757309 - 767244 on Build GRCh38 | Deletion | WNK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |