Assay Details
Target Gene Details
Entrez Gene ID: | 161 |
Gene Name: | adaptor related protein complex 2 alpha 2 subunit |
Gene Aliases: |
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ |
Location: |
Chr.11:925809-1012245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A2 | NM_001242837.1 | NP_001229766.1 | ||
NM_012305.3 | NP_036437.1 | |||
XM_011519929.1 | XP_011518231.1 | |||
XM_011519930.2 | XP_011518232.1 | |||
AB020706.2 | BAA74922.2 | |||
AK001986.1 | ||||
AK027891.1 | BAB55435.1 | |||
AK223558.1 | BAD97278.1 | |||
AK293763.1 | ||||
AK299364.1 | ||||
BC006155.1 | AAH06155.1 | |||
BG705558.1 | ||||
BI462785.1 | ||||
DB461570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625088 | Chr.11:876861 - 979340 on Build GRCh38 | Gain |
![]() ![]() |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2667274 | Chr.11:935793 - 940156 on Build GRCh38 | Deletion |
![]() |
dgv175e214 | Chr.11:851409 - 1043855 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv428244 | Chr.11:882364 - 1061595 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)