Assay Details
Target Gene Details
Entrez Gene ID: | 10522 |
Gene Name: | DEAF1, transcription factor |
Gene Aliases: |
MRD24, NUDR, SPN, ZMYND5 |
Location: |
Chr.11:644220-695754 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DEAF1 | NM_001293634.1 | NP_001280563.1 | ||
NM_021008.3 | NP_066288.2 | |||
XM_011519842.2 | XP_011518144.1 | |||
AB209831.1 | BAD93068.1 | |||
AF007165.1 | AAB62704.1 | |||
AF049459.1 | AAC79676.1 | |||
AF049460.1 | AAC79677.1 | |||
AF068892.1 | AAC25714.1 | |||
AF068893.1 | AAC25715.1 | |||
AF068894.1 | AAC25716.1 | |||
AF068895.1 | AAC25717.1 | |||
AF068896.1 | AAC25718.1 | |||
AF068897.1 | AAC25719.1 | |||
AK289873.1 | ||||
AK291383.1 | ||||
BC053322.1 | AAH53322.1 | |||
FJ985253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2743861 | Chr.11:664235 - 664895 on Build GRCh38 | Deletion |
![]() |
dgv124e201 | Chr.11:664307 - 665458 on Build GRCh38 | Deletion |
![]() |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1039558 | Chr.11:534242 - 672091 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1053964 | Chr.11:491334 - 692433 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1525n54 | Chr.11:647869 - 688312 on Build GRCh38 | Loss |
![]() |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv543e59 | Chr.11:664352 - 665150 on Build GRCh38 | Duplication |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)