Assay Details
Target Gene Details
Entrez Gene ID: | 3703 |
Gene Name: | STT3A, catalytic subunit of the oligosaccharyltransferase complex |
Gene Aliases: |
ITM1, STT3-A, TMC |
Location: |
Chr.11:125592795-125622759 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STT3A | NM_001278503.1 | NP_001265432.1 | ||
NM_001278504.1 | NP_001265433.1 | |||
NM_152713.4 | NP_689926.1 | |||
XM_011542807.2 | XP_011541109.1 | |||
AK130548.1 | ||||
AK290040.1 | ||||
AK290657.1 | ||||
AK295892.1 | ||||
BC020965.2 | AAH20965.1 | |||
BC048348.1 | ||||
BT007100.1 | AAP35764.1 | |||
BX649102.1 | ||||
L38961.1 | AAB05994.1 | |||
L47337.1 | AAL77539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)