Assay Details
Target Gene Details
Entrez Gene ID: | 29974 |
Gene Name: | APOBEC1 complementation factor |
Gene Aliases: |
ACF, ACF64, ACF65, APOBEC1CF, ASP |
Location: |
Chr.10:50799409-50885681 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A1CF | NM_001198818.1 | NP_001185747.1 | ||
NM_001198819.1 | NP_001185748.1 | |||
NM_001198820.1 | NP_001185749.1 | |||
NM_014576.3 | NP_055391.2 | |||
NM_138932.2 | NP_620310.1 | |||
NM_138933.2 | NP_620311.1 | |||
XM_005269718.2 | XP_005269775.1 | |||
XM_005269720.3 | XP_005269777.1 | |||
XM_011539729.2 | XP_011538031.1 | |||
XM_011539730.2 | XP_011538032.1 | |||
XM_017016160.1 | XP_016871649.1 | |||
AF209192.1 | AAF34824.1 | |||
AF271789.1 | AAF76221.1 | |||
AF271790.1 | AAF76222.1 | |||
AJ272078.1 | CAB94754.1 | |||
AJ272079.1 | CAB94755.1 | |||
AK000324.1 | BAA91086.1 | |||
AK291982.1 | ||||
AK303801.1 | ||||
BC130519.1 | ||||
BC144196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035197 | Chr.10:50686822 - 50862589 on Build GRCh38 | Gain | ASAH2B A1CF LOC729047 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |