Assay Details
Target Gene Details
Entrez Gene ID: | 23071 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
PDIA10, TXNDC4 |
Location: |
Chr.9:99979179-100099052 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERP44 | NM_015051.2 | NP_055866.1 | ||
AB011145.1 | ||||
AJ344330.2 | CAC87611.1 | |||
AK075024.1 | ||||
AY359048.1 | AAQ89407.1 | |||
BC005374.1 | AAH05374.1 | |||
BX648519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1050510 | Chr.9:99922036 - 99986376 on Build GRCh38 | Gain | ERP44 STX17 |
nsv831668 | Chr.9:99805887 - 99985011 on Build GRCh38 | Gain+Loss | STX17-AS1 ERP44 STX17 NR4A3 LOC101928438 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |