Assay Details
Target Gene Details
Entrez Gene ID: | 1955 |
Gene Name: | multiple EGF like domains 9 |
Gene Aliases: |
EGFL5 |
Location: |
Chr.9:120600918-120714487 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEGF9 | NM_001080497.2 | NP_001073966.2 | ||
AB011542.1 | BAA32470.1 | |||
AK295380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33143 | Chr.9:120491423 - 120708811 on Build GRCh38 | Loss | CDK5RAP2 MEGF9 |
nsv831711 | Chr.9:120503030 - 120705363 on Build GRCh38 | Gain | CDK5RAP2 MEGF9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |