Assay Details
Target Gene Details
Entrez Gene ID: | 727 |
Gene Name: | complement component 5 |
Gene Aliases: |
C5D, C5a, C5b, CPAMD4, ECLZB |
Location: |
Chr.9:120952335-121075174 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C5 | NM_001317163.1 | NP_001304092.1 | ||
NM_001735.2 | NP_001726.2 | |||
XM_011518980.2 | XP_011517282.1 | |||
XM_017015102.1 | XP_016870591.1 | |||
XM_017015103.1 | XP_016870592.1 | |||
AB209031.1 | BAD92268.1 | |||
BC113738.1 | ||||
BC113740.1 | ||||
M57729.1 | AAA51925.1 | |||
M65134.1 | AAA51856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759713 | Chr.9:120885919 - 121080717 on Build GRCh38 | Loss | PHF19 C5 CNTRL TRAF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |