Assay Details
Target Gene Details
Entrez Gene ID: | 100506753 |
Gene Name: | AZIN1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:102864271-102977876 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AZIN1-AS1 | NR_126338.1 | |||
NR_126339.1 | ||||
BF939259.1 | ||||
CD675847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611822 | Chr.8:102804878 - 102882098 on Build GRCh38 | Loss | LOC107986873 AZIN1 AZIN1-AS1 LOC401472 |
esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss | KLF10 UBR5 LOC107986873 ODF1 ATP6V1C1 AZIN1 LOC101927245 AZIN1-AS1 LOC401472 |