Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | GTF2IP7 TMEM120A STYXL1 SNORA14A POR LOC107986711 MDH2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | HIP1 ZP3 GTF2IP7 SRRM3 PMS2P10 POR POMZP3 LOC105375355 LOC541473 CCL24 PMS2P3 LOC441259 TRIM73 HSPB1 POM121C SPDYE5 YWHAG CCL26 SSC4D GTF2IRD2B SPDYE16 MIR4651 LOC100133091 NCF1C MDH2 GTF2IP1 FDPSP2 DTX2 TMEM120A STAG3L1 STYXL1 SNORA14A PMS2P2 LOC107986711 NSUN5P1 RHBDD2 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | TMEM120A STYXL1 SNORA14A POR MIR4651 |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | HIP1 ZP3 GTF2IP7 SRRM3 POR POMZP3 LOC105375355 CCL24 PMS2P3 HSPB1 POM121C SPDYE5 YWHAG CCL26 SSC4D SPDYE16 MIR4651 LOC100133091 MDH2 FDPSP2 DTX2 TMEM120A STYXL1 SNORA14A LOC107986711 RHBDD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |