Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | HSPB1 SPDYE16 POMZP3 LOC100133091 YWHAG FDPSP2 ZP3 LOC105375355 SSC4D SRRM3 DTX2 DTX2P1-UPK3BP1-PMS2P11 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | SPDYE16 CCDC146 POMZP3 LOC100133091 YWHAG ZP3 FGL2 DTX2P1-UPK3BP1-PMS2P11 SPDYE17 SPDYE18 FDPSP2 LOC105375355 PMS2P9 SSC4D DTX2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | RHBDD2 GTF2IP1 POMZP3 CCL24 TMEM120A LOC100133091 MIR4651 GTF2IP7 NSUN5P1 STAG3L1 POR STYXL1 HIP1 PMS2P3 HSPB1 CCL26 LOC107986711 SPDYE16 NCF1C TRIM73 YWHAG ZP3 SPDYE5 PMS2P2 LOC441259 POM121C SNORA14A LOC541473 GTF2IRD2B FDPSP2 MDH2 LOC105375355 SSC4D SRRM3 PMS2P10 DTX2 |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | YWHAG ZP3 SSC4D |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | RHBDD2 POMZP3 CCL24 TMEM120A LOC100133091 MIR4651 GTF2IP7 POR STYXL1 HIP1 PMS2P3 HSPB1 CCL26 LOC107986711 SPDYE16 YWHAG ZP3 SPDYE5 POM121C SNORA14A FDPSP2 MDH2 LOC105375355 SSC4D SRRM3 DTX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |