Assay Details
Target Gene Details
Entrez Gene ID: | 4698 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A5 |
Gene Aliases: |
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13 |
Location: |
Chr.7:123536997-123601651 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA5 | NM_001282419.2 | NP_001269348.1 | ||
NM_001282420.2 | NP_001269349.1 | |||
NM_001282421.2 | NP_001269350.1 | |||
NM_001282422.2 | NP_001269351.1 | |||
NM_001291304.1 | NP_001278233.1 | |||
NM_005000.4 | NP_004991.1 | |||
NR_104168.2 | ||||
NR_104169.2 | ||||
NR_111925.1 | ||||
NR_111926.1 | ||||
AK315458.1 | ||||
AV713284.1 | ||||
BC000813.1 | AAH00813.1 | |||
BC020821.1 | AAH20821.1 | |||
BC070236.1 | AAH70236.1 | |||
BC070237.1 | AAH70237.1 | |||
BF671339.1 | ||||
BF790429.1 | ||||
BG400546.1 | ||||
BG611486.1 | ||||
BT006695.1 | AAP35341.1 | |||
CR933664.1 | CAI45962.1 | |||
U53468.1 | AAB02224.1 | |||
U64028.1 | AAB37259.1 | |||
W44870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3614856 | Chr.7:123545800 - 123551524 on Build GRCh38 | Loss | NDUFA5 |
nsv831116 | Chr.7:123447558 - 123619922 on Build GRCh38 | Gain+Loss | LOC102724555 NDUFA5 ASB15 IQUB |
nsv524192 | Chr.7:122763367 - 123578468 on Build GRCh38 | Gain | SLC13A1 TAS2R16 NDUFA5 ASB15 CADPS2 IQUB |
esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain | SLC13A1 RNF148 TAS2R16 RNF133 WASL HYAL4 LOC102724555 HYALP1 NDUFA5 ASB15 AASS FEZF1 PTPRZ1 LMOD2 CADPS2 IQUB FEZF1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |