Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AB051763.1 | BAB18572.1 | |||
AB209874.1 | BAD93111.1 | |||
AF258341.1 | AAG09798.1 | |||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BC034277.1 | AAH34277.1 | |||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 | ||||
S90469.1 | AAB21814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | STYXL1 POR GTF2IP7 SNORA14A TMEM120A LOC107986711 MDH2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SPDYE5 LOC441259 POR SPDYE16 NSUN5P1 FDPSP2 SRRM3 STYXL1 PMS2P3 CCL26 SNORA14A DTX2 GTF2IP1 LOC107986711 HSPB1 STAG3L1 PMS2P2 LOC100133091 CCL24 HIP1 RHBDD2 TRIM73 TMEM120A NCF1C YWHAG LOC541473 GTF2IRD2B LOC105375355 GTF2IP7 MIR4651 PMS2P10 SSC4D POM121C POMZP3 ZP3 MDH2 |
esv3613746 | Chr.7:75965095 - 76003876 on Build GRCh38 | Loss | STYXL1 POR TMEM120A |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | STYXL1 POR SNORA14A MIR4651 TMEM120A |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE5 POR SPDYE16 FDPSP2 SRRM3 STYXL1 PMS2P3 CCL26 SNORA14A DTX2 LOC107986711 HSPB1 LOC100133091 CCL24 HIP1 RHBDD2 TMEM120A YWHAG LOC105375355 GTF2IP7 MIR4651 SSC4D POM121C POMZP3 ZP3 MDH2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |