Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | LOC105375355 DTX2P1-UPK3BP1-PMS2P11 ZP3 SPDYE16 YWHAG FDPSP2 SSC4D POMZP3 LOC100133091 SRRM3 HSPB1 DTX2 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | LOC105375355 DTX2P1-UPK3BP1-PMS2P11 ZP3 SPDYE16 FDPSP2 POMZP3 FGL2 CCDC146 DTX2 SPDYE17 SPDYE18 YWHAG SSC4D LOC100133091 PMS2P9 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | TMEM120A LOC105375355 ZP3 SPDYE16 FDPSP2 PMS2P3 TRIM73 STYXL1 LOC441259 RHBDD2 GTF2IRD2B CCL26 LOC107986711 SSC4D CCL24 LOC100133091 HSPB1 POM121C SNORA14A SPDYE5 NCF1C POMZP3 HIP1 GTF2IP7 MIR4651 DTX2 STAG3L1 NSUN5P1 POR MDH2 PMS2P10 YWHAG PMS2P2 LOC541473 SRRM3 GTF2IP1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | TMEM120A LOC105375355 ZP3 SPDYE16 FDPSP2 PMS2P3 STYXL1 RHBDD2 CCL26 LOC107986711 SSC4D CCL24 LOC100133091 HSPB1 POM121C SNORA14A SPDYE5 POMZP3 HIP1 GTF2IP7 MIR4651 DTX2 POR MDH2 YWHAG SRRM3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |