Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | POR LOC107986711 GTF2IP7 SNORA14A MDH2 STYXL1 TMEM120A |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SRRM3 LOC541473 DTX2 HSPB1 LOC107986711 MDH2 YWHAG FDPSP2 PMS2P10 LOC100133091 SSC4D GTF2IRD2B CCL24 LOC105375355 SPDYE5 POR RHBDD2 POM121C PMS2P2 SNORA14A STYXL1 LOC441259 ZP3 TRIM73 MIR4651 NSUN5P1 STAG3L1 NCF1C CCL26 PMS2P3 HIP1 GTF2IP7 SPDYE16 GTF2IP1 TMEM120A POMZP3 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 POR SNORA14A STYXL1 TMEM120A |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SRRM3 DTX2 HSPB1 LOC107986711 MDH2 YWHAG FDPSP2 LOC100133091 SSC4D CCL24 LOC105375355 SPDYE5 POR RHBDD2 POM121C SNORA14A STYXL1 ZP3 MIR4651 CCL26 PMS2P3 HIP1 GTF2IP7 SPDYE16 TMEM120A POMZP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |