Assay Details
Target Gene Details
Entrez Gene ID: | 90523 |
Gene Name: | muscular LMNA-interacting protein |
Gene Aliases: |
C6orf142 |
Location: |
Chr.6:54018916-54266325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLIP | NM_001281746.1 | NP_001268675.1 | ||
NM_138569.2 | NP_612636.2 | |||
XM_005249476.4 | XP_005249533.1 | |||
XM_005249478.4 | XP_005249535.1 | |||
XM_005249480.4 | XP_005249537.1 | |||
AK055530.1 | BAB70942.1 | |||
BX647567.1 | ||||
DA761851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759431 | Chr.6:53974575 - 54142551 on Build GRCh38 | Gain+Loss | LOC101927189 MLIP MLIP-IT1 |
nsv523301 | Chr.6:54031357 - 54056329 on Build GRCh38 | Loss | MLIP |
nsv1018062 | Chr.6:53977790 - 54060394 on Build GRCh38 | Gain | LOC101927189 MLIP MLIP-IT1 |
nsv603110 | Chr.6:54021120 - 54238996 on Build GRCh38 | Gain | MLIP |
nsv5307 | Chr.6:54009320 - 54087432 on Build GRCh38 | Deletion | MLIP |
nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain | KLHL31 LOC101927189 LRRC1 LINC01564 MLIP MLIP-IT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |