Assay Details
Target Gene Details
Entrez Gene ID: | 1235 |
Gene Name: | C-C motif chemokine receptor 6 |
Gene Aliases: |
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22 |
Location: |
Chr.6:167111807-167139141 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCR6 | NM_004367.5 | NP_004358.2 | ||
NM_031409.3 | NP_113597.2 | |||
AK223494.1 | BAD97214.1 | |||
AK293015.1 | ||||
BC037960.1 | AAH37960.1 | |||
CD692892.1 | ||||
DA645096.1 | ||||
DN996219.1 | ||||
U68030.1 | AAC51124.1 | |||
U68031.1 | AAD14874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752099 | Chr.6:167034693 - 167165614 on Build GRCh38 | Gain | LOC105378123 FGFR1OP GPR31 CCR6 |
nsv469673 | Chr.6:167086888 - 167245044 on Build GRCh38 | Loss | LOC105378123 TCP10L2 GPR31 CCR6 |
nsv1024657 | Chr.6:167112710 - 167142000 on Build GRCh38 | Loss | CCR6 |
esv2759492 | Chr.6:167087716 - 167776053 on Build GRCh38 | Gain | UNC93A LOC105378123 LOC441178 TTLL2 LOC105378127 TCP10 LOC401286 TCP10L2 LOC105378130 GPR31 CCR6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |