Assay Details
Target Gene Details
Entrez Gene ID: | 100874282 |
Gene Name: | MLIP intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.6:53998890-54007152 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLIP-IT1 | NR_046832.1 | |||
CF890740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759431 | Chr.6:53974575 - 54142551 on Build GRCh38 | Gain+Loss | MLIP MLIP-IT1 LOC101927189 |
nsv1018062 | Chr.6:53977790 - 54060394 on Build GRCh38 | Gain | MLIP MLIP-IT1 LOC101927189 |
nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain | KLHL31 LRRC1 MLIP LINC01564 MLIP-IT1 LOC101927189 |