Assay Details
Target Gene Details
Entrez Gene ID: | 166752 |
Gene Name: | FRAS1 related extracellular matrix 3 |
Gene Aliases: |
- |
Location: |
Chr.4:143577407-143700675 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FREM3 | NM_001168235.1 | NP_001161707.1 | ||
AI184955.1 | ||||
DR731220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759288 | Chr.4:143427140 - 144210439 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830103 | Chr.4:143472245 - 143631892 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3894047 | Chr.4:143572032 - 144009062 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)