Assay Details
Target Gene Details
Entrez Gene ID: | 57619 |
Gene Name: | shroom family member 3 |
Gene Aliases: |
APXL3, MSTP013, SHRM, ShrmL |
Location: |
Chr.4:76435100-76783253 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHROOM3 | NM_020859.3 | NP_065910.3 | ||
XM_011532158.2 | XP_011530460.1 | |||
XM_011532159.1 | XP_011530461.1 | |||
AB055660.1 | BAB84689.1 | |||
AK055294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759262 | Chr.4:76343443 - 76534252 on Build GRCh38 | Loss | SHROOM3 CCDC158 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |