Assay Details
Target Gene Details
Entrez Gene ID: | 55209 |
Gene Name: | SET domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.3:9397700-9478154 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv277n21 | Chr.3:9390066 - 9453435 on Build GRCh38 | Loss | THUMPD3-AS1 SETD5 |