Assay Details
Target Gene Details
Entrez Gene ID: | 100131635 |
Gene Name: | hCG1645011-like |
Gene Aliases: |
- |
Location: |
Chr.3:187702366-187732415 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100131635 | NR_034062.1 | |||
AW235368.1 |
Target Gene Details
Entrez Gene ID: | 344892 |
Gene Name: | receptor transporter protein 2 |
Gene Aliases: |
Z3CXXC2 |
Location: |
Chr.3:187698259-187715625 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RTP2 | XM_017006301.1 | XP_016861790.1 | ||
XM_017006302.1 | XP_016861791.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)