Assay Details
Target Gene Details
Entrez Gene ID: | 2257 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
FGF12B, FHF1 |
Location: |
Chr.3:192139393-192727599 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF12 | NM_004113.5 | NP_004104.3 | ||
NM_021032.4 | NP_066360.1 | |||
XM_005247227.2 | XP_005247284.1 | |||
XM_006713538.3 | XP_006713601.1 | |||
XM_017005879.1 | XP_016861368.1 | |||
AK125307.1 | ||||
AK312513.1 | ||||
AK313671.1 | ||||
BC022524.1 | AAH22524.1 | |||
DA521146.1 | ||||
U66197.1 | AAB18913.1 | |||
U76381.2 | AAB18786.3 | |||
Z70276.1 | CAA94240.1 |
Target Gene Details
Entrez Gene ID: | 100873986 |
Gene Name: | FGF12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:192238630-192283097 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF12-AS1 | NR_046596.1 | |||
DB034608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2726410 | Chr.3:192219587 - 192270877 on Build GRCh38 | Deletion |
![]() ![]() |
esv3599140 | Chr.3:192218451 - 192270327 on Build GRCh38 | Gain |
![]() ![]() |
nsv435755 | Chr.3:192218493 - 192272921 on Build GRCh38 | Deletion |
![]() ![]() |
nsv4170 | Chr.3:192180843 - 192272191 on Build GRCh38 | Deletion |
![]() ![]() |
esv3569197 | Chr.3:192259290 - 192298866 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)