Assay Details
Target Gene Details
Entrez Gene ID: | 253152 |
Gene Name: | epoxide hydrolase 4 |
Gene Aliases: |
ABHD7, EH4, EPHXRP |
Location: |
Chr.1:92029976-92063536 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHX4 | NM_173567.4 | NP_775838.3 | ||
AK074822.1 | BAC11230.1 | |||
BC041475.1 | AAH41475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002730 | Chr.1:91836984 - 92099672 on Build GRCh38 | Gain | EPHX4 SETSIP TGFBR3 BRDT BTBD8 |
nsv524845 | Chr.1:91750168 - 92133148 on Build GRCh38 | Gain | EPHX4 SETSIP TGFBR3 BRDT BTBD8 |
nsv830581 | Chr.1:91851650 - 92045618 on Build GRCh38 | Loss | EPHX4 TGFBR3 BRDT |
esv2750842 | Chr.1:91891224 - 92114118 on Build GRCh38 | Gain | EPHX4 SETSIP TGFBR3 BRDT BTBD8 |
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion | SNORA66 HSP90B3P CDC7 HFM1 SETSIP TGFBR3 FAM69A EVI5 SNORD21 BTBD8 RPL5 EPHX4 KIAA1107 GLMN GFI1 C1orf146 BRDT RPAP2 |
nsv1006318 | Chr.1:91750851 - 92142767 on Build GRCh38 | Gain | EPHX4 SETSIP TGFBR3 BRDT BTBD8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |