Assay Details
Target Gene Details
Entrez Gene ID: | 1380 |
Gene Name: | complement component 3d receptor 2 |
Gene Aliases: |
C3DR, CD21, CR, CVID7, SLEB9 |
Location: |
Chr.1:207454300-207489895 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CR2 | NM_001006658.2 | NP_001006659.1 | ||
NM_001877.4 | NP_001868.2 | |||
XM_011509206.2 | XP_011507508.1 | |||
AK223627.1 | BAD97347.1 | |||
AK301496.1 | ||||
AU133057.1 | ||||
BC090937.1 | AAH90937.1 | |||
BC136394.1 | ||||
J03565.1 | AAA35784.1 | |||
M26004.1 | AAA35786.1 | |||
Y00649.1 | CAA68674.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758993 | Chr.1:207384550 - 207751722 on Build GRCh38 | Gain+Loss | CR1L CR2 CR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |