Assay Details
Target Gene Details
Entrez Gene ID: | 554203 |
Gene Name: | JPX transcript, XIST activator (non-protein coding) |
Gene Aliases: |
DCBALD06, ENOX, LINC00183, NCRNA00183 |
Location: |
Chr.X:73944324-74070384 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JPX | NR_024582.1 | |||
AK056172.1 | ||||
BP871721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29793 | Chr.X:73945896 - 73949840 on Build GRCh38 | Loss |
![]() |
esv1005511 | Chr.X:73941104 - 73966080 on Build GRCh38 | Insertion |
![]() |
esv2656629 | Chr.X:73946227 - 73949977 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)