Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | MIR6857 MAGED2 USP51 FGD1 ALAS2 MAGEH1 TRO MIR4536-2 GPR173 MIRLET7F2 WNK3 HUWE1 TSPYL2 MIR4536-1 GNL3L XAGE3 TSR2 FAM104B IQSEC2 SNORA11 MIR6894 PAGE3 HSD17B10 MIR6895 RIBC1 FOXR2 SMC1A MTRNR2L10 KANTR FAM156B LOC100421746 APEX2 SNORA109 PAGE5 ITIH6 FAM120C FAM156A MIR98 PFKFB1 PAGE2 KDM5C PAGE2B PHF8 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | MIR6857 MAGED2 USP51 FGD1 ALAS2 MAGEH1 TRO MIR4536-2 GPR173 MIRLET7F2 WNK3 HUWE1 TSPYL2 MIR4536-1 GNL3L XAGE3 TSR2 FAM104B IQSEC2 SNORA11 MIR6894 PAGE3 HSD17B10 MIR6895 RIBC1 FOXR2 SMC1A MTRNR2L10 KANTR FAM156B LOC100421746 APEX2 SNORA109 PAGE5 ITIH6 FAM120C FAM156A MIR98 PFKFB1 PAGE2 KDM5C PAGE2B PHF8 |