Assay Details
Target Gene Details
Entrez Gene ID: | 10134 |
Gene Name: | B-cell receptor-associated protein 31 |
Gene Aliases: |
6C6-AG, BAP31, CDM, DDCH, DXS1357E |
Location: |
Chr.X:153700492-153724746 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAP31 | NM_001139441.1 | NP_001132913.1 | ||
NM_001139457.2 | NP_001132929.1 | |||
NM_001256447.1 | NP_001243376.1 | |||
NM_005745.7 | NP_005736.3 | |||
AK057613.1 | ||||
AK125631.1 | ||||
AK222494.1 | BAD96214.1 | |||
AK223059.1 | BAD96779.1 | |||
AU132745.1 | ||||
BC014323.1 | AAH14323.1 | |||
BC065292.1 | AAH65292.1 | |||
BE729820.1 | ||||
BX474419.1 | ||||
CX164660.1 | ||||
X81109.1 | CAA57015.1 | |||
Z31696.1 | CAA83501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | MIR718 OPN1MW IDH3G BGN HCFC1-AS1 MIR3202-1 NAA10 PDZD4 ARHGAP4 UBL4A MIR3202-2 SSR4 MECP2 FAM223A L1CAM ZFP92 DUSP9 DNASE1L1 TEX28 TMEM187 FLNA OPN1LW ATP2B3 LAGE3 HCFC1 BCAP31 LOC105373383 TREX2 CTAG1A MIR6858 MAGEA1 LOC107983954 ATP6AP1 FAM58A OPN1MW3 PLXNB3 FAM3A EMD SNORA70 LOC105373377 SLC10A3 SRPK3 PNCK HAUS7 OPN1MW2 G6PD PLXNA3 ABCD1 ZNF275 LCA10 IKBKG LOC105373378 SLC6A8 AVPR2 LOC107987332 PNMA6A RPL10 FAM50A GDI1 CH17-340M24.3 LOC105373386 TAZ LOC105373381 IRAK1 TKTL1 RENBP |
esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss | PNCK SLC6A8 BCAP31 ABCD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |