Assay Details
Target Gene Details
Entrez Gene ID: | 401612 |
Gene Name: | solute carrier family 25 member 53 |
Gene Aliases: |
MCART6 |
Location: |
Chr.X:104099214-104157027 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A53 | NM_001012755.4 | NP_001012773.2 | ||
AL833609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469772 | Chr.X:103928672 - 104131003 on Build GRCh38 | Gain | H2BFXP SLC25A53 H2BFM ZCCHC18 LOC286437 H2BFWT LOC100101478 TMSB15B |
esv2758880 | Chr.X:103827471 - 104210380 on Build GRCh38 | Gain | FAM199X H2BFXP SLC25A53 H2BFM ZCCHC18 LL0XNC01-116E7.2 LOC286437 H2BFWT LOC100101478 RAB9B TMSB15B |
nsv519626 | Chr.X:103933493 - 104158390 on Build GRCh38 | Gain | FAM199X H2BFXP SLC25A53 H2BFM ZCCHC18 LOC286437 H2BFWT LOC100101478 TMSB15B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |