Assay Details
Target Gene Details
Entrez Gene ID: | 100302692 |
Gene Name: | FTX transcript, XIST regulator (non-protein coding) |
Gene Aliases: |
LINC00182, MIR374AHG, NCRNA00182 |
Location: |
Chr.X:74028136-74293574 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FTX | NR_028379.1 | |||
AK311345.1 |
Target Gene Details
Entrez Gene ID: | 554203 |
Gene Name: | JPX transcript, XIST activator (non-protein coding) |
Gene Aliases: |
DCBALD06, ENOX, LINC00183, NCRNA00183 |
Location: |
Chr.X:73944324-74070384 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JPX | NR_024582.1 |