Assay Details
Target Gene Details
Entrez Gene ID: | 5475 |
Gene Name: | protein phosphatase with EF-hand domain 1 |
Gene Aliases: |
PP7, PPEF, PPP7C, PPP7CA |
Location: |
Chr.X:18675909-18827921 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPEF1 | NM_006240.2 | NP_006231.2 | ||
NM_152224.1 | NP_689410.1 | |||
NM_152226.1 | NP_689412.1 | |||
XM_005274553.2 | XP_005274610.1 | |||
XM_017029612.1 | XP_016885101.1 | |||
AK292235.1 | ||||
BC036026.1 | AAH36026.1 | |||
X97867.1 | CAA66461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33517 | Chr.X:18661048 - 19128069 on Build GRCh38 | Loss | PPEF1-AS1 LOC101928415 PHKA2-AS1 PHKA2 PPEF1 ADGRG2 RS1 |
esv2752317 | Chr.X:18691267 - 19292303 on Build GRCh38 | Gain | PPEF1-AS1 LOC101928415 PHKA2-AS1 PHKA2 PPEF1 ADGRG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |