Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | CTAG1B SLC10A3 OPN1MW MIR664B FAM50A RPL10 HCFC1-AS1 FAM223B DKC1 GAB3 LOC105373386 LOC107987332 MIR718 LOC101927830 H2AFB2 FAM3A GDI1 MIR1184-1 SNORA70 IRAK1 SNORA36A CLIC2 F8A3 MECP2 OPN1MW2 MIR3202-2 CTAG1A F8 UBL4A F8A1 VBP1 MIR1184-3 ATP6AP1 G6PD TAZ MPP1 SNORA56 CH17-340M24.3 OPN1LW FUNDC2 CMC4 TKTL1 DNASE1L1 SPRY3 MTCP1 F8A2 EMD BRCC3 TMLHE HCFC1 MIR6858 PLXNA3 MIR1184-2 H2AFB1 OPN1MW3 TEX28 MIR3202-1 RAB39B CTAG2 H2AFB3 IKBKG TMEM187 FAM223A FLNA LAGE3 SMIM9 TMLHE-AS1 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | SPRY3 TMLHE |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | SPRY3 TMLHE |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | MIR1184-1 RAB39B FUNDC2 CLIC2 CMC4 F8A3 H2AFB3 F8 F8A1 SPRY3 VBP1 MTCP1 MIR1184-3 F8A2 BRCC3 TMLHE LOC101927830 MIR1184-2 H2AFB1 SMIM9 MPP1 TMLHE-AS1 H2AFB2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |