Assay Details
Target Gene Details
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv21578 | Chr.X:49251822 - 49253132 on Build GRCh38 | Gain |
![]() |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)