Assay Details
Target Gene Details
Entrez Gene ID: | 89801 |
Gene Name: | protein phosphatase 1 regulatory subunit 3F |
Gene Aliases: |
HB2E, LL0XNC01-7P3.1, R3F |
Location: |
Chr.X:49269822-49301465 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R3F | NM_001184745.1 | NP_001171674.1 | ||
NM_033215.4 | NP_149992.3 | |||
XM_005272687.3 | XP_005272744.1 | |||
XM_017029932.1 | XP_016885421.1 | |||
XM_017029933.1 | XP_016885422.1 | |||
XM_017029934.1 | XP_016885423.1 | |||
XM_017029935.1 | XP_016885424.1 | |||
AK056909.1 | ||||
AK303738.1 | ||||
BC131588.1 | ||||
DC412626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | GAGE10 FOXP3 CACNA1F PPP1R3F CCDC22 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX6 PRICKLE3 FOXP3 HDAC6 CACNA1F PORCN GLOD5 GRIPAP1 SSX5 PCSK1N PIM2 ERAS SPACA5 WDR13 SSX9 PQBP1 RBM3 KCND1 SLC35A2 ZNF630-AS1 SLC38A5 CCDC120 OTUD5 WDR45 GATA1 MAGIX TIMM17B SPACA5B FTSJ1 PRAF2 SYP-AS1 SUV39H1 PPP1R3F PLP2 TFE3 SSX4 ZNF630 SSX3 GPKOW SYP SSX4B EBP ZNF182 SSX1 LOC101927635 WAS CCDC22 TBC1D25 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | GAGE10 FOXP3 CACNA1F PPP1R3F CCDC22 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |