Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | TRO FAM156B PFKFB1 SNORA11 GPR173 APEX2 MIR4536-1 IQSEC2 WNK3 PHF8 FAM156A PAGE2 HSD17B10 FAM120C PAGE5 TSR2 ITIH6 SNORA109 MIR98 SMC1A PAGE2B KDM5C FAM104B TSPYL2 MIR6857 MAGEH1 MIR6895 RIBC1 USP51 KANTR FGD1 MIR6894 MAGED2 MIR4536-2 ALAS2 MIRLET7F2 XAGE3 MTRNR2L10 FOXR2 PAGE3 LOC100421746 HUWE1 GNL3L |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | TRO FAM156B PFKFB1 SNORA11 GPR173 APEX2 MIR4536-1 IQSEC2 WNK3 PHF8 FAM156A PAGE2 HSD17B10 FAM120C PAGE5 TSR2 ITIH6 SNORA109 MIR98 SMC1A PAGE2B KDM5C FAM104B TSPYL2 MIR6857 MAGEH1 MIR6895 RIBC1 USP51 KANTR FGD1 MIR6894 MAGED2 MIR4536-2 ALAS2 MIRLET7F2 XAGE3 MTRNR2L10 FOXR2 PAGE3 LOC100421746 HUWE1 GNL3L |