Assay Details
Target Gene Details
Entrez Gene ID: | 54328 |
Gene Name: | G protein-coupled receptor 173 |
Gene Aliases: |
SREB3 |
Location: |
Chr.X:53049324-53080615 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR173 | NM_018969.5 | NP_061842.1 | ||
BC009861.2 | AAH09861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508773 | Chr.X:53034408 - 53141410 on Build GRCh38 | Insertion | GPR173 KANTR TSPYL2 |
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | GNL3L WNK3 PAGE3 SNORA109 PAGE5 PAGE2B FAM120C MIR6857 MAGEH1 PAGE2 MIR4536-2 MIRLET7F2 HUWE1 APEX2 HSD17B10 MIR6894 PFKFB1 FGD1 IQSEC2 ITIH6 FAM156B MIR6895 USP51 ALAS2 MAGED2 SMC1A TRO TSR2 MIR4536-1 FOXR2 LOC100421746 KANTR PHF8 XAGE3 MIR98 FAM156A SNORA11 GPR173 MTRNR2L10 FAM104B RIBC1 KDM5C TSPYL2 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | GNL3L WNK3 PAGE3 SNORA109 PAGE5 PAGE2B FAM120C MIR6857 MAGEH1 PAGE2 MIR4536-2 MIRLET7F2 HUWE1 APEX2 HSD17B10 MIR6894 PFKFB1 FGD1 IQSEC2 ITIH6 FAM156B MIR6895 USP51 ALAS2 MAGED2 SMC1A TRO TSR2 MIR4536-1 FOXR2 LOC100421746 KANTR PHF8 XAGE3 MIR98 FAM156A SNORA11 GPR173 MTRNR2L10 FAM104B RIBC1 KDM5C TSPYL2 |
esv3576878 | Chr.X:52894443 - 53071885 on Build GRCh38 | Gain | FAM156A GPR173 FAM156B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |