Assay Details
Target Gene Details
Entrez Gene ID: | 5063 |
Gene Name: | p21 (RAC1) activated kinase 3 |
Gene Aliases: |
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK |
Location: |
Chr.X:110944241-111227361 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAK3 | NM_001128166.2 | NP_001121638.1 | ||
NM_001128167.2 | NP_001121639.1 | |||
NM_001324325.1 | NP_001311254.1 | |||
NM_001324330.1 | NP_001311259.1 | |||
XM_011530968.2 | XP_011529270.1 | |||
XM_011530969.1 | XP_011529271.1 | |||
XM_017029557.1 | XP_016885046.1 | |||
XM_017029559.1 | XP_016885048.1 | |||
XM_017029561.1 | XP_016885050.1 | |||
AK290504.1 | ||||
AK315284.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |