Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SPRY3 TEX28 TMEM187 CH17-340M24.3 H2AFB2 ATP6AP1 LOC107987332 SNORA56 TAZ TMLHE MIR1184-2 VBP1 TMLHE-AS1 LAGE3 MIR664B DNASE1L1 CTAG2 MIR718 FAM223B MIR3202-2 CTAG1B SNORA36A RAB39B MIR3202-1 SMIM9 FUNDC2 H2AFB3 F8A1 G6PD CTAG1A BRCC3 F8A3 SNORA70 IRAK1 UBL4A LOC105373386 CLIC2 MIR1184-3 OPN1MW TKTL1 MPP1 F8A2 FAM3A OPN1MW3 FAM50A FAM223A MIR1184-1 MIR6858 LOC101927830 HCFC1-AS1 OPN1MW2 F8 MECP2 FLNA PLXNA3 CMC4 GDI1 GAB3 RPL10 MTCP1 IKBKG SLC10A3 OPN1LW DKC1 H2AFB1 EMD HCFC1 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | SPRY3 TMLHE |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | SPRY3 TMLHE |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | MIR1184-3 SPRY3 F8 H2AFB2 MPP1 RAB39B F8A2 CMC4 SMIM9 FUNDC2 H2AFB3 F8A1 MIR1184-1 TMLHE MTCP1 BRCC3 F8A3 MIR1184-2 VBP1 TMLHE-AS1 LOC101927830 H2AFB1 CLIC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |