Assay Details
Target Gene Details
Entrez Gene ID: | 1183 |
Gene Name: | chloride voltage-gated channel 4 |
Gene Aliases: |
CLC4, ClC-4, ClC-4A |
Location: |
Chr.X:10156945-10237660 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLCN4 | NM_001256944.1 | NP_001243873.1 | ||
NM_001830.3 | NP_001821.2 | |||
AB019432.1 | BAA77327.1 | |||
AK225287.1 | ||||
AK289564.1 | ||||
AK299611.1 | ||||
BC036068.1 | AAH36068.1 | |||
BC130278.1 | ||||
BP310547.1 | ||||
X77197.1 | CAA54417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758861 | Chr.X:10083096 - 10263971 on Build GRCh38 | Loss | WWC3 CLCN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |