Assay Details
Target Gene Details
Entrez Gene ID: | 54328 |
Gene Name: | G protein-coupled receptor 173 |
Gene Aliases: |
SREB3 |
Location: |
Chr.X:53049324-53080615 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR173 | NM_018969.5 | NP_061842.1 | ||
BC009861.2 | AAH09861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508773 | Chr.X:53034408 - 53141410 on Build GRCh38 | Insertion | TSPYL2 KANTR GPR173 |
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | GNL3L PAGE2B PFKFB1 FAM104B SNORA109 KANTR PAGE3 WNK3 APEX2 MIR4536-1 PHF8 MAGED2 HSD17B10 GPR173 FAM156A KDM5C USP51 MTRNR2L10 XAGE3 MAGEH1 MIR6894 ITIH6 SNORA11 FAM120C HUWE1 FAM156B TSPYL2 TSR2 IQSEC2 MIR6895 FOXR2 MIR6857 MIRLET7F2 ALAS2 TRO MIR98 LOC100421746 PAGE2 PAGE5 SMC1A RIBC1 MIR4536-2 FGD1 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | GNL3L PAGE2B PFKFB1 FAM104B SNORA109 KANTR PAGE3 WNK3 APEX2 MIR4536-1 PHF8 MAGED2 HSD17B10 GPR173 FAM156A KDM5C USP51 MTRNR2L10 XAGE3 MAGEH1 MIR6894 ITIH6 SNORA11 FAM120C HUWE1 FAM156B TSPYL2 TSR2 IQSEC2 MIR6895 FOXR2 MIR6857 MIRLET7F2 ALAS2 TRO MIR98 LOC100421746 PAGE2 PAGE5 SMC1A RIBC1 MIR4536-2 FGD1 |
esv3576878 | Chr.X:52894443 - 53071885 on Build GRCh38 | Gain | FAM156B FAM156A GPR173 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |