Assay Details
Target Gene Details
Entrez Gene ID: | 3358 |
Gene Name: | 5-hydroxytryptamine receptor 2C |
Gene Aliases: |
5-HT1C, 5-HT2C, 5-HTR2C, 5HTR2C, HTR1C |
Location: |
Chr.X:114584078-114910061 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR2C | NM_000868.3 | NP_000859.1 | ||
NM_001256760.2 | NP_001243689.1 | |||
NM_001256761.2 | NP_001243690.1 | |||
AK295753.1 | ||||
AK308766.1 | ||||
M81778.1 | AAA60317.1 | |||
U49516.1 | AAB40898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3574320 | Chr.X:114607663 - 114619561 on Build GRCh38 | Loss | HTR2C |
nsv527347 | Chr.X:114537428 - 114983201 on Build GRCh38 | Gain | SNORA35 MIR1264 MIR1912 MIR448 MIR1298 MIR1911 HTR2C MIR764 |
esv33994 | Chr.X:113685854 - 115738426 on Build GRCh38 | Gain+Loss | SNORA35 MIR448 PLS3-AS1 MIR1911 IL13RA2 HTR2C MIR764 RBMXL3 PLS3 LRCH2 MIR1264 MIR1912 MIR1298 LUZP4 LOC105373314 |
dgv524n21 | Chr.X:114067803 - 114619628 on Build GRCh38 | Gain | HTR2C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |